missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ TBE Gels, 10%

Novex™ TBE Gels 10% designed to run on the XCell SureLock™ Mini-Cell provide high-resolution analysis of restriction digests and PCR products.

Brand:  Invitrogen™ EC62755BOX

126.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 12045735

  • £180.00 / Pack of 10
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: Catalog numbers for gels refer to a single box. One box contains 10 gels.
This item is not returnable. View return policy

Description

Description

Novex™ TBE Gels 10% provide high-resolution analysis of restriction digests and PCR products. Designed to run on the XCell SureLock™ Mini-Cell, the polyacrylamide gels give sharp, clearly resolved, intense bands, and provide separations of double-strand DNA fragments from 35–120 bp.

Novex TBE Gels are available in a variety of well formats and gel percentages and can be stained by silver staining, ethidium­ bromide, and SYBR™ Green staining techniques after electrophoresis.

Advantages of TBE Gels for nucleic acid separation:
• High resolution and sensitivity with lower background staining
• Requires ∼10% sample concentration and volume of large or agarose gels
• Efficient blotting
• Easy extraction of DNA from gels
• Gel extraction does not interfere with enzymatic reactions
• Accurate and reproducible results

Formulation
Novex TBE gels are manufactured with high-purity Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS.

Recommended buffers
For optimum performance, Novex™ TBE Running Buffer and Novex™ Hi-Density TBE Sample Buffer are strongly recommended for use with these gels. Novex Hi-Density TBE Sample Buffer contains the tracking dyes Bromophenol Blue and Xylene Cyanol as well as the density agent Ficoll™, which yields sharper and straighter bands than conventional density agents.

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Catalog number refers to a single box of 10 gels

Store at 4°C.
XCell SureLock Mini-Cell
TBE
35 to 120 bp
10 Gels/Box
Wet Ice
Novex TBE Gels, 10%, 15-well
10%
TBE Gel
15-well
Double-stranded DNA (dsDNA)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.