Learn More
Invitrogen™ Novex™ TBE Gels, 10%
Novex™ TBE Gels 10% designed to run on the XCell SureLock™ Mini-Cell provide high-resolution analysis of restriction digests and PCR products.
£180.00 - £183.00
Specifications
Content And Storage | • Catalog number refers to a single box of 10 gels Store at 4°C. |
---|---|
For Use With (Equipment) | XCell SureLock Mini-Cell |
Gel Percentage | 10% |
Gel Type | TBE |
Product Type | TBE Gel |
Product Code | Brand | Description | Wells | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Wells | Price | Quantity & Availability | ||||
12025735
|
Invitrogen™
EC6275BOX |
Novex TBE Gels, 10%, 10-well | 10-well |
£182.00
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
12035735
|
Invitrogen™
EC62752BOX |
Novex TBE Gels, 10%, 12-well | 12-well |
£183.00
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
12045735
|
Invitrogen™
EC62755BOX |
Novex TBE Gels, 10%, 15-well | 15-well |
£180.00
Pack of 10 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
Description
Novex™ TBE Gels 10% provide high-resolution analysis of restriction digests and PCR products. Designed to run on the XCell SureLock™ Mini-Cell, the polyacrylamide gels give sharp, clearly resolved, intense bands, and provide separations of double-strand DNA fragments from 35–120 bp.
Novex TBE Gels are available in a variety of well formats and gel percentages and can be stained by silver staining, ethidium bromide, and SYBR™ Green staining techniques after electrophoresis.
Advantages of TBE Gels for nucleic acid separation:
• High resolution and sensitivity with lower background staining
• Requires ∼10% sample concentration and volume of large or agarose gels
• Efficient blotting
• Easy extraction of DNA from gels
• Gel extraction does not interfere with enzymatic reactions
• Accurate and reproducible results
Formulation
Novex TBE gels are manufactured with high-purity Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS.
Recommended buffers
For optimum performance, Novex™ TBE Running Buffer and Novex™ Hi-Density TBE Sample Buffer are strongly recommended for use with these gels. Novex Hi-Density TBE Sample Buffer contains the tracking dyes Bromophenol Blue and Xylene Cyanol as well as the density agent Ficoll™, which yields sharper and straighter bands than conventional density agents.
Order Info
Shipping Condition: Wet Ice
Specifications
• Catalog number refers to a single box of 10 gels Store at 4°C. |
|
10% | |
TBE Gel | |
10 Gels/Box | |
Wet Ice |
XCell SureLock Mini-Cell | |
TBE | |
35 to 120 bp | |
Double-stranded DNA (dsDNA) |