missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELISA TMB Stabilized Chromogen
For use with all sandwich ELISAs which utilize Horseradish Peroxidase (HRP) as detecting enzyme
£43.66 - £284.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10523405
|
Invitrogen™
SB01 |
25 mL |
£43.66
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10354603
|
Invitrogen™
SB02 |
1 L |
£284.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Formulation: Liquid
- Available in two sizes
- Detection method: Colormetric
Build Your Own Immunoassay, Cell Analysis, Immunoassays
Order Info
Shipping Condition: Wet Ice
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ ELISA TMB Stabilized Chromogen