missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELISA TMB Stabilized Chromogen

For use with all sandwich ELISAs which utilize Horseradish Peroxidase (HRP) as detecting enzyme

Brand:  Invitrogen™ SB02

246.64 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 10354603

  • £284.00 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • Formulation: Liquid
  • Available in two sizes
  • Detection method: Colormetric

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 L
Solution. Store at 2-8°C.
Colorimetric
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number77581001 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberSB02
Lot Number77197917 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberSB02
Lot Number77197935 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberSB02
Lot Number77312205 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberSB02
Lot Number77328561 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberSB02
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

    Days

    37

    Hours

    12

    Minutes

    59

    Seconds

    51
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ ELISA TMB Stabilized Chromogen > 1L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.