missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tricine Welcome Pack, 16%

The Novex Tricine Welcome Pack provides all of the necessary Novex Tricine gels, buffers, and reagents you will need to begin using the Mini Gel Tank.

Brand:  Invitrogen™ EC6695C

Additional Details : Weight : 2.60900kg

 View more versions of this product

Product Code. 17124809

  • £692.00 / Each
Estimated Shipment: 29-07-2024
to see stock.
Add to basket

Includes:
  • Mini Gel Tank
  • Novex Tricine Mini Gels (2 boxes, 20 gels)
  • Novex Tricine SDS Running Buffer (10x)
  • Novex Tricine SDS Sample Buffer (2X)
  • NuPAGE Sample Reducing Agent (10X)
  • Spectra Multicolor Low Range Protein Ladder

Description

Description

The Novex Tricine Welcome Pack provides all of the necessary Novex Tricine gels, buffers, and reagents you will need to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies being used.

The Novex Tricine Welcome Pack contains:
• Mini Gel Tank (A25977)
• Novex Tricine Mini Gels (2 boxes, 20 gels)
• Novex Tricine SDS Running Buffer, 10X (LC1675)
• Novex Tricine SDS Sample Buffer, 2X (LC1676)
• NuPAGE Sample Reducing Agent, 10X (NP0009)
• Spectra Multicolor Low Range Protein Ladder (26628)

About the Mini Gel Tank
The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies being used.

About Novex Tricine gels
Novex Tricine gels provide separation of low molecular weight proteins and peptides. The Tricine system is a modification of the tris-glycine discontinuous buffer system developed by Schaegger and von Jagow specifically for peptides and low molecular weight proteins. In this system the tricine replaces the glycine in the running buffer, resulting in more efficient stacking and de-stacking of low molecular weight proteins and higher resolution of smaller peptides.

Transfer
For transfer of proteins to a membrane, we recommend using Novex Tris-Glycine Transfer Buffer (LC3675) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Specifications

Specifications

Mini Gel Tank
Mini
Tricine
15-well
1 Welcome Pk.
16%
1.0 mm
2.5 to 200 kDa
Novex™
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers