Learn More
Tocris Bioscience™ NSC 23766
Selective inhibitor of Rac1-GEF interaction; antioncogenic
Brand:  Tocris Bioscience™ 2161/50
471.00 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18014632
  • £600.00 / 50mg
Explore more special offers
Description
Description
NSC 23766 is a selective inhibitor of Rac1-GEF interaction. NSC 23766 prevents Rac1 activation by Rac-specific guanine nucleotide exchange factors (GEFs) TrioN and Tiam1 (IC50 ∼ 50 μM) without affecting Cdc42 or RhoA activation. The compound inhibits Rac1-mediated cell functions and is reported to reverse tumor cell phenotypes in prostate cancer cells. NSC 23766 also inhibits mammary carcinoma cell invasion in vitro and inhibits mammary tumor progression and metastasis in a mouse model.
Specifications
Specifications
>98%
NSC 23766
Rho GTPase Inhibitors
50 mg
530.97