Learn More
NSC319726, MedChemExpress
MedChemExpress NSC319726 (ZMC1) is a mutant p53R175 reactivator; inhibits growth of fibroblasts expressing the p53R175 mutation (IC50 = 8 nM); shows no inhibition for p53 wild-type cells.
Brand: Medchem Express HY-18634
This item is not returnable.
View return policy
Description
NSC319726 (ZMC1) is a mutant p53R175 reactivatorSpecifications
NSC319726 | |
Off-White | |
C11H14N4S | |
ZMC1 | |
C/C(C1=NC=CC=C1)=N\NC(N2CCC2)=S | |
234.32 | |
Research | |
Cancer-programmed cell death |
71555-25-4 | |
Research | |
50 mg | |
DMSO : 31.25 mg/mL (133.36 mM; Need ultrasonic) | |
234.32 | |
98.06% | |
Powder |