Learn More
Tocris Bioscience™ NTRC 824
Potent and selective non-peptide NTS2 antagonist
Brand:  Tocris Bioscience™ 5438/50
775.79 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18736509
  • £1010.00 / 50mg
Explore more special offers
Description
Description
NTRC 824 is a potent and selective non-peptide neurotensin (NTS) type-2 receptor antagonist (IC50 = 38 nM). Exhibits >150-fold selectivity for NTS2 over NTS1.
Specifications
Specifications
N-[2-[5-[[(4-Methylphenyl)sulfonyl]amino]-3-(2,2,2-trifluoroacetyl)-1H-indol-1-yl]acetyl]-L-leucine
50 mg
C25H26F3N3O6S
>98%
1623002-61-8
Neurotensin Receptor Antagonists
553.55