missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NTRC 824

Potent and selective non-peptide NTS2 antagonist

£259.00 - £1010.00

Specifications

Chemical Name or Material N-[2-[5-[[(4-Methylphenyl)sulfonyl]amino]-3-(2,2,2-trifluoroacetyl)-1H-indol-1-yl]acetyl]-L-leucine
CAS 1623002-61-8
Target Neurotensin Receptor Antagonists
Molecular Formula C25H26F3N3O6S
Formula Weight 553.55
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18798698
View Documents
Tocris Bioscience™
5438/10
10 mg
£259.00
10mg
Please to purchase this item. Need a web account? Register with us today!
18736509
View Documents
Tocris Bioscience™
5438/50
50 mg
£1010.00
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent and selective non-peptide neurotensin (NTS) type-2 receptor antagonist (IC50 = 38 nM). Exhibits >150-fold selectivity for NTS2 over NTS1.
Specifications

Specifications

N-[2-[5-[[(4-Methylphenyl)sulfonyl]amino]-3-(2,2,2-trifluoroacetyl)-1H-indol-1-yl]acetyl]-L-leucine
Neurotensin Receptor Antagonists
553.55
1623002-61-8
C25H26F3N3O6S
>98%
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.