missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ Xtra Maxi Kits
Purify transfection-grade plasmid DNA with high yield. The NucleoBond™ Xtra Maxi Kit represents the new generation of anion exchanger. The optimized design results in high yield and ultra-fast processing.
£397.00 - £2100.12
Specifications
Buffer | Yes |
---|---|
Format | Maxi Gravity-flow Column |
Isolation Technology | Anion Exchange Chromatography Technology |
RNase | Yes |
Sample Volume | <600mL (high copy), <1200mL (low copy) |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11902312
|
Macherey-Nagel™
740414.100 |
100 |
£2100.12
100 preps |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12792603
|
Macherey-Nagel™
740414.10 |
10 |
£397.00
0.10kg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12748412
|
Macherey-Nagel™
740414.50 |
50 |
£1820.00
50 preps |
In Stock
Log in to see stock. |
|||||
Description
Features
- Optimized column design—Silica resin packed between filters, with prep in just 75 minutes
- Column filter included—High filter flow rates, parallel lysate clearing, and loading onto the column
- Improved silica material—Typical yields of 1,000μg and can be used over a wide pH range
- Proven anion-exchange technology—Transfection-grade plasmid DNA
- Kits contain: NucleoBond™ Xtra Maxi columns and filter, RNase A, buffers (RES, LYS, NEU, EQU, WASH, ELU) and plastic washers
Applications
- High-copy plasmids from E. coli
- Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications
Yes | |
Anion Exchange Chromatography Technology | |
<600mL (high copy), <1200mL (low copy) | |
75 min./prep |
Maxi Gravity-flow Column | |
Yes | |
<300kbp | |
1000μg |