Learn More
Macherey-Nagel™ Nucleodur™ C18 Isis Analytical HPLC EC Column
£448.00 - £635.00
Specifications
Column Chemistry | Silica |
---|---|
Column Format | Analytical Column |
Crosslinking | Polymeric octadecyl |
For Use With | HPLC |
Mobile Phase Compatibility | Acetonitrile/water |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
12468447
|
Macherey-Nagel™
760412.20 |
2 mm | 125 mm |
£520.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12408457
|
Macherey-Nagel™
760413.20 |
2 mm | 150 mm |
£520.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12478447
|
Macherey-Nagel™
760412.30 |
3 mm | 125 mm |
£525.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12458457
|
Macherey-Nagel™
760414.30 |
3 mm | 250 mm |
£600.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12448457
|
Macherey-Nagel™
760414.20 |
2 mm | 250 mm |
£605.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12418457
|
Macherey-Nagel™
760413.30 |
3 mm | 150 mm |
£525.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12428447
|
Macherey-Nagel™
760410.20 |
2 mm | 50 mm |
£448.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12438447
|
Macherey-Nagel™
760410.30 |
3 mm | 50 mm |
£448.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12478457
|
Macherey-Nagel™
760415.46 |
4.6 mm | 100 mm |
£580.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12498447
|
Macherey-Nagel™
760412.46 |
4.6 mm | 125 mm |
£580.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12438457
|
Macherey-Nagel™
760413.46 |
4.6 mm | 150 mm |
£580.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12468457
|
Macherey-Nagel™
760414.46 |
4.6 mm | 250 mm |
£635.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12767830
|
Macherey-Nagel™
760416.46 |
4.6 mm | 75 mm |
£491.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12458447
|
Macherey-Nagel™
760410.46 |
4.6 mm | 50 mm |
£491.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
10406964
|
Macherey-Nagel™
760414.40 |
4 mm | 250 mm |
£600.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12428457
|
Macherey-Nagel™
760413.40 |
4 mm | 150 mm |
£525.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12448447
|
Macherey-Nagel™
760410.40 |
4 mm | 50 mm |
£448.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
Specifications
Silica | |
Polymeric octadecyl | |
Acetonitrile/water | |
1 to 10 | |
1 Ea. | |
L1 | |
Silica | |
Steric Selectivity | |
Analytical EC Column |
Analytical Column | |
HPLC | |
5 μm | |
110 Å | |
Acetonitrile/water | |
C18 Isis | |
20% | |
Yes |