missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NUCLEODUR™ HILIC Column for HPLC, 3μm Particles
Ideal for analytical HPLC applications with hydrophilic compounds. Macherey-Nagel™ NUCLEODUR™ HILIC Column for HPLC, 3μm Particles is an EC standard column with high purity silica phase. The columns are available in a variety of diameters and lengths.
Brand: Macherey-Nagel™ 760534.46
This item is not returnable.
View return policy
Description
This product is suitable for Hydrophilic Interaction Chromatography (HILIC) applications. It features a highly pure ammonium/sulfonic acid-modified silica phase with an 80:20 acetonitrile-water eluent. These EC standard columns have a particle size of 3μm and are used in analytical HPLC.
- Ideal for reproducible and stable chromatography of highly polar analytes
- Suitable for analytical and preparative applications
- Very short column conditioning period
- Used with hydrophilic compounds, such as: organic polar acids and bases, polar natural compounds, nucleosides, oligonucleotides, amino acids, peptides, water soluble vitamins
Specifications
Ammonium /sulfonic acid | |
Ammonium /sulfonic acid | |
0.8 mL/min | |
Hydrophobic | |
100 mm | |
80:20 Acetonitrile/water | |
3 μm | |
110 Å | |
1 Ea. | |
340 m²/g | |
Silica | |
HILIC | |
HILIC | |
EC Column |
Analytical Column | |
4.6 mm | |
For Reproducible and Stable Chromatography of Highly Polar Analytes | |
Yes | |
8700 psi (600 bar) | |
Spherical | |
2 to 8.5 | |
0.9 mL/g | |
80:20 Acetonitrile/water | |
Ammonium/Sulfonic Acid Modified Silica | |
7% | |
No | |
Stainless Steel |