Learn More
Macherey-Nagel™ NUCLEODUR™ HILIC Column for HPLC, 3μm Particles
Ideal for analytical HPLC applications with hydrophilic compounds. Macherey-Nagel™ NUCLEODUR™ HILIC Column for HPLC, 3μm Particles is an EC standard column with high purity silica phase. The columns are available in a variety of diameters and lengths.
£601.00 - £823.00
Specifications
Column Chemistry Ammonium /sulfonic acid
Column Format Analytical Column
Crosslinking Ammonium /sulfonic acid
For Use With For Reproducible and Stable Chromatography of Highly Polar Analytes
Functionality/Interaction Type Hydrophobic
View More Specs
Products 15
Product Code Brand Length (Metric) Diameter (Metric) Flow Rate Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Flow Rate Price Quantity & Availability  
10668614
View Documents
Macherey-Nagel™
760534.20
100 mm 2 mm 0.15 mL/min
£684.00
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
10311625
View Documents
Macherey-Nagel™
760531.20
125 mm 2 mm 0.15 mL/min
£684.00
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
10711325
View Documents
Macherey-Nagel™
760530.20
250 mm 2 mm 0.15 mL/min
£793.00
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
12738862
View Documents
Macherey-Nagel™
760532.20
50 mm 2 mm 0.15 mL/min
£601.00
Each
In Stock
to see stock.
Add to basket
10480864
View Documents
Macherey-Nagel™
760531.30
125 mm 3 mm 0.34 mL/min
£694.00
Each
In Stock
to see stock.
Add to basket
12728862
View Documents
Macherey-Nagel™
760530.30
250 mm 3 mm 0.34 mL/min
£787.00
Each
In Stock
to see stock.
Add to basket
12717850
View Documents
Macherey-Nagel™
760534.46
100 mm 4.6 mm 0.8 mL/min
£746.00
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
12748862
View Documents
Macherey-Nagel™
760532.30
50 mm 3 mm 0.34 mL/min
£601.00
Each
In Stock
to see stock.
Add to basket
12408467
View Documents
Macherey-Nagel™
760531.46
125 mm 4.6 mm 0.8 mL/min
£746.00
Each
In Stock
to see stock.
Add to basket
12428467
View Documents
Macherey-Nagel™
760533.46
150 mm 4.6 mm 0.8 mL/min
£746.00
Each
In Stock
to see stock.
Add to basket
12488457
View Documents
Macherey-Nagel™
760530.46
250 mm 4.6 mm 0.8 mL/min
£823.00
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
12707850
View Documents
Macherey-Nagel™
760532.46
50 mm 4.6 mm 0.8 mL/min
£653.00
Each
In Stock
to see stock.
Add to basket
10627715
View Documents
Macherey-Nagel™
760530.40
250 mm 4 mm 0.6 mL/min
£787.00
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
12498457
View Documents
Macherey-Nagel™
760531.40
125 mm 4 mm 0.6 mL/min
£694.00
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
12418467
View Documents
Macherey-Nagel™
760532.40
50 mm 4 mm 0.6 mL/min
£601.00
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
Description
Description
This product is suitable for Hydrophilic Interaction Chromatography (HILIC) applications. It features a highly pure ammonium/sulfonic acid-modified silica phase with an 80:20 acetonitrile-water eluent. These EC standard columns have a particle size of 3μm and are used in analytical HPLC.
  • Ideal for reproducible and stable chromatography of highly polar analytes
  • Suitable for analytical and preparative applications
  • Very short column conditioning period
  • Used with hydrophilic compounds, such as: organic polar acids and bases, polar natural compounds, nucleosides, oligonucleotides, amino acids, peptides, water soluble vitamins
Specifications
Specifications
Ammonium /sulfonic acid
Ammonium /sulfonic acid
Hydrophobic
8700 psi (600 bar)
Spherical
2 to 8.5
0.9 mL/g
80:20 Acetonitrile/water
Ammonium/Sulfonic Acid Modified Silica
7%
No
Stainless Steel
Analytical Column
For Reproducible and Stable Chromatography of Highly Polar Analytes
Yes
80:20 Acetonitrile/water
3 μm
110 Å
1 Ea.
340 m²/g
Silica
HILIC
HILIC
EC Column