missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NUCLEODUR™ HILIC Column for HPLC, 3μm Particles

Ideal for analytical HPLC applications with hydrophilic compounds. Macherey-Nagel™ NUCLEODUR™ HILIC Column for HPLC, 3μm Particles is an EC standard column with high purity silica phase. The columns are available in a variety of diameters and lengths.

Brand:  Macherey-Nagel™ 760532.30

 View more versions of this product

Product Code. 12748862

  • £601.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This product is suitable for Hydrophilic Interaction Chromatography (HILIC) applications. It features a highly pure ammonium/sulfonic acid-modified silica phase with an 80:20 acetonitrile-water eluent. These EC standard columns have a particle size of 3μm and are used in analytical HPLC.

  • Ideal for reproducible and stable chromatography of highly polar analytes
  • Suitable for analytical and preparative applications
  • Very short column conditioning period
  • Used with hydrophilic compounds, such as: organic polar acids and bases, polar natural compounds, nucleosides, oligonucleotides, amino acids, peptides, water soluble vitamins
Specifications

Specifications

Ammonium /sulfonic acid
Ammonium /sulfonic acid
0.34 mL/min
Hydrophobic
50 mm
80:20 Acetonitrile/water
3 μm
110 Å
1 Ea.
340 m²/g
Ammonium/Sulfonic Acid Modified Silica
7%
No
Stainless Steel
Analytical Column
3 mm
For Reproducible and Stable Chromatography of Highly Polar Analytes
Yes
8700 psi (600 bar)
Spherical
2 to 8.5
0.9 mL/g
80:20 Acetonitrile/water
60°C
Silica
HILIC
HILIC
EC Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers