missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoMag™ NGS Clean-up and Size Select
NucleoMag™ NGS Beads Suspension that enable magnetic bead-based clean-up and size selection of NGS library preparation reactions for use in Next Gen Sequencing.
Brand: Macherey-Nagel™ 744970.500
4162.90 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
NucleoMag™ NGS Clean-up and Size Select
- Clean-up and size selection steps in NGS library kits
- Sample input as little as 17.5 pg
- Sample volume of 50 μL to 150 μL
- Tunable size selection 150-800 bp
- Convenient magnetic bead technology
- Easy to adjust for specific applications or sequencers
- Optimal scalability for manual and automated processing
Specifications
Refrigerated at 4 - 8 °C | |
Reaction Mixtures from Common NGS Library Kits | |
Magnetic separator; Elution and separation plates | |
NucleoMag NGS-Beads in buffer | |
17.5 pg to 5 μg, Input volume: 50 - 150μL | |
Highly Reactive Superparamagnetic Beads |
Magnetic Bead | |
Elution volume: 10 - 100 μL | |
NGS clean-up and size selection | |
500 mL | |
Typical recovery: ≥ 80 % |