missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoMag™ NGS Clean-up and Size Select
NucleoMag™ NGS Beads Suspension that enable magnetic bead-based clean-up and size selection of NGS library preparation reactions for use in Next Gen Sequencing.
£125.00 - £4525.00
Specifications
Content And Storage | Refrigerated at 4 - 8 °C |
---|---|
Isolation Technology | Magnetic Bead |
Sample Type | Reaction Mixtures from Common NGS Library Kits |
Elution Volume | Elution volume: 10 - 100 μL |
For Use With (Equipment) | Magnetic separator; Elution and separation plates |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15889167
|
Macherey-Nagel™
744970.5 |
5 mL |
£125.00
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15899167
|
Macherey-Nagel™
744970.50 |
50 mL |
£819.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15809177
|
Macherey-Nagel™
744970.500 |
500 mL |
£4525.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NucleoMag™ NGS Clean-up and Size Select
- Clean-up and size selection steps in NGS library kits
- Sample input as little as 17.5 pg
- Sample volume of 50 μL to 150 μL
- Tunable size selection 150-800 bp
- Convenient magnetic bead technology
- Easy to adjust for specific applications or sequencers
- Optimal scalability for manual and automated processing
Specifications
Refrigerated at 4 - 8 °C | |
Reaction Mixtures from Common NGS Library Kits | |
Magnetic separator; Elution and separation plates | |
NucleoMag NGS-Beads in buffer | |
Typical recovery: ≥ 80 % |
Magnetic Bead | |
Elution volume: 10 - 100 μL | |
NGS clean-up and size selection | |
17.5 pg to 5 μg, Input volume: 50 - 150μL | |
Highly Reactive Superparamagnetic Beads |
Safety and Handling
missing translation for 'shelfLife' : Up to 6 months