Learn More
Cytiva Nucleon™ PhytoPure™ Genomic DNA Extraction Kits
Nucleon PhytoPure DNA Extraction kits use a phytopure resin to bind plant polysaccharides ensuring high yields of high-quality plant genomic DNA in only one hour
Brand:  Cytiva RPN8511
Code : GH
Additional Details : CAS Number : 9048-46-8 Weight : 0.52000kg
 View more versions of this product
Product Code. 10410875
  • £447.00 / 50 reactions
Estimated Shipment: 06-08-2024
to see stock.
Description
Description
  • Rapid protocol does not require phenol or CTAB
  • Two kit sizes provide economical scale-up using the same phenol-free protocol
  • Effective removal of polysaccharides minimises inhibition of downstream applications
Specifications
Specifications
Quality, length, and purity of a phenol-chloroform extraction without phenol. Nucleon PhytoPure enables rapid chloroform extraction of high-quality, high molecular weight genomic DNA from plant and fungal samples with efficient removal of protein as well
Successfully extracts DNA from many plant species, starting with fresh, frozen, or freeze dried material
Nucleon PhytoPure
Barley (Fresh, Frozen or Freeze Dried), Maize (Fresh, Frozen or Freeze Dried), Rye (Fresh, Frozen or Freeze Dried), Wheat (Fresh, Frozen or Freeze Dried)
Reagents 1 and 2 and PhytoPure™ Resin
1000 mg