missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Nucleon™ PhytoPure™ Genomic DNA Extraction Kits
Nucleon PhytoPure DNA Extraction kits use a phytopure resin to bind plant polysaccharides ensuring high yields of high-quality plant genomic DNA in only one hour
£234.00 - £447.00
Specifications
Description | Quality, length, and purity of a phenol-chloroform extraction without phenol. Nucleon PhytoPure enables rapid chloroform extraction of high-quality, high molecular weight genomic DNA from plant and fungal samples with efficient removal of protein as well |
---|---|
Sample Type | Barley (Fresh, Frozen or Freeze Dried), Maize (Fresh, Frozen or Freeze Dried), Rye (Fresh, Frozen or Freeze Dried), Wheat (Fresh, Frozen or Freeze Dried) |
For Use With (Application) | Successfully extracts DNA from many plant species, starting with fresh, frozen, or freeze dried material |
Includes | Reagents 1 and 2 and PhytoPure™ Resin |
Includes: Reagents 1 and 2 and PhytoPure Resin
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10762037
|
Cytiva
RPN8510 |
100 mg |
£234.00
50 reactions |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10410875
|
Cytiva
RPN8511 |
1000 mg |
£447.00
50 reactions |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Rapid protocol does not require phenol or CTAB
- Two kit sizes provide economical scale-up using the same phenol-free protocol
- Effective removal of polysaccharides minimises inhibition of downstream applications
Specifications
Quality, length, and purity of a phenol-chloroform extraction without phenol. Nucleon PhytoPure enables rapid chloroform extraction of high-quality, high molecular weight genomic DNA from plant and fungal samples with efficient removal of protein as well | |
Barley (Fresh, Frozen or Freeze Dried), Maize (Fresh, Frozen or Freeze Dried), Rye (Fresh, Frozen or Freeze Dried), Wheat (Fresh, Frozen or Freeze Dried) | |
Successfully extracts DNA from many plant species, starting with fresh, frozen, or freeze dried material | |
Reagents 1 and 2 and PhytoPure™ Resin |