Learn More
Cytiva Nucleon™ PhytoPure™ Genomic DNA Extraction Kits
Nucleon PhytoPure DNA Extraction kits use a phytopure resin to bind plant polysaccharides ensuring high yields of high-quality plant genomic DNA in only one hour
Brand: Cytiva RPN8510
216.22 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : GH
Additional Details : CAS Number : 9048-46-8 Weight : 0.20300kg
Description
- Rapid protocol does not require phenol or CTAB
- Two kit sizes provide economical scale-up using the same phenol-free protocol
- Effective removal of polysaccharides minimises inhibition of downstream applications
Specifications
Quality, length, and purity of a phenol-chloroform extraction without phenol. Nucleon PhytoPure enables rapid chloroform extraction of high-quality, high molecular weight genomic DNA from plant and fungal samples with efficient removal of protein as well | |
Successfully extracts DNA from many plant species, starting with fresh, frozen, or freeze dried material | |
illustra Nucleon PhytoPure |
Barley (Fresh, Frozen or Freeze Dried), Maize (Fresh, Frozen or Freeze Dried), Rye (Fresh, Frozen or Freeze Dried), Wheat (Fresh, Frozen or Freeze Dried) | |
Reagents 1 and 2 and PhytoPure™ Resin | |
100 mg |