Learn More
Macherey-Nagel™ Nucleosil™ Standard C18 CC Guard Columns
Brand:  Macherey-Nagel™ 721783.40
 View more versions of this product
Product Code. 12742073
  • £153.00 / Pack of 3
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
11%
5 μm
120Å
4 mm
Acetonitrile/Water
Silica
Guard Column
HPLC
Yes
Yes
2 to 8
8 mm
HPLC CC Guard Column
NUCLEOSIL™ Standard C18
Acetonitrile/Water
Octadecyl
RP chromatography
Spherical
Hydrophobic
L1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Nucleosil™ Standard C18 CC Guard Columns > Pore Size: 120A