Learn More
Macherey-Nagel™ NucleoSpin™ 96 DNA Plasma
High throughput solution for isolation of cell-free circulating DNA from up to 2 mL blood plasma in 96-well format. Available in full or core kits, with one or four plates.
Brand: Macherey-Nagel™ 740873.1
Additional Details : Weight : 1.35000kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Description
NucleoSpin™ 96 DNA Plasma Kits
- High throughput solution for cell-free DNA isolation
- Fragmented DNA of 50 bp and larger can be purified with high efficiency
- Manual or automated processing
- Protocol follows state-of-the-art bind-wash-elute procedures
- Optimized for Cell-Free DNA BCT™ ('Streck Tubes'), or for standard EDTA blood draw tubes
- Core Kits include basic content focused on automation platforms
- Typical downstream applications: NGS, qPCR, ddPCR
Specifications
Room temperature (18-25 °C) | |
Silica Membrane Technology | |
Elution volume: 100 μL | |
Isolation of cell-free, circulating DNA from plasma | |
1 x 96 | |
0.5-2 mL | |
Depends on sample source, storage, and quality |
96-well Plate | |
Blood Plasma (esp. human) | |
Vacuum manifold/pump | |
NucleoSpin Plasma Binding Plate, 96-Square-well Block, Round-well Block, buffers, Proteinase K | |
Full Kit | |
90 min/96 preps |