missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ 96 DNA Plasma
High throughput solution for isolation of cell-free circulating DNA from up to 2 mL blood plasma in 96-well format. Available in full or core kits, with one or four plates.
£348.00 - £1350.00
Specifications
Content And Storage | Room temperature (18-25 °C) |
---|---|
Format | 96-well Plate |
Isolation Technology | Silica Membrane Technology |
Sample Type | Blood Plasma (esp. human) |
Elution Volume | Elution volume: 100 μL |
Product Code | Brand | No. of Samples | Product Type | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Samples | Product Type | Includes | Price | Quantity & Availability | |||
15688455
|
Macherey-Nagel™
740874.1 |
1 x 96 | Core Kit | NucleoSpin Plasma Binding Plate, buffers, Proteinase K |
£348.00
1 set |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15698455
|
Macherey-Nagel™
740874.4 |
4 x 96 | Core Kit | NucleoSpin Plasma Binding Plate, buffers, Proteinase K |
£1155.00
4 sets |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15668455
|
Macherey-Nagel™
740873.1 |
1 x 96 | Full Kit | NucleoSpin Plasma Binding Plate, 96-Square-well Block, Round-well Block, buffers, Proteinase K |
£525.00
1 set |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15678455
|
Macherey-Nagel™
740873.4 |
4 x 96 | Full Kit | NucleoSpin Plasma Binding Plate, 96-Square-well Block, Round-well Block, buffers, Proteinase K |
£1350.00
4 sets |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
NucleoSpin™ 96 DNA Plasma Kits
- High throughput solution for cell-free DNA isolation
- Fragmented DNA of 50 bp and larger can be purified with high efficiency
- Manual or automated processing
- Protocol follows state-of-the-art bind-wash-elute procedures
- Optimized for Cell-Free DNA BCT™ ('Streck Tubes'), or for standard EDTA blood draw tubes
- Core Kits include basic content focused on automation platforms
- Typical downstream applications: NGS, qPCR, ddPCR
Specifications
Room temperature (18-25 °C) | |
Silica Membrane Technology | |
Elution volume: 100 μL | |
Isolation of cell-free, circulating DNA from plasma | |
90 min/96 preps |
96-well Plate | |
Blood Plasma (esp. human) | |
Vacuum manifold/pump | |
0.5-2 mL | |
Depends on sample source, storage, and quality |