missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Nucleospin™ 96 Plasmid Transfection-grade

A fast way to purify transfection-grade plasmid DNA using mini-format technology to reduce endotoxins from bacterial lysates. Full and Core kits utilize a 96-well plate format for an efficient high-throughput process.

Brand:  Macherey-Nagel™ 740492.4

1018.13 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.



 View more versions of this product

Product Code. 15553636

  • £1315.00 / 384 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Endotoxins are co-purified during typical plasmid preparations from bacterial lysates. Since they interfere with eukaryotic cell survival, endotoxin reduction is essential prior to cell transfection. NucleoSpin™ Plasmid Transfection-grade kits are designed to reduce endotoxins from bacterial lysates.

  • Purifies high-copy plasmids for transfection
  • Novel technology to diminish endotoxin content (< 50 EU/μg DNA) for successful transfection
  • Rapid purification from E. coli via automatic, manual, vacuum or centrifugation processing
  • Ultra-fast procedure simplifies the work-flow
  • High-throughput — 96-well plates allow for easy automation and supports plasmid screening
  • Downstream applications: transfection of common cell lines (lipofection, electroporation)
  • Also suitable for standard molecular biology application, such as cloning and sequencing
Specifications

Specifications

<50 EU/ng DNA
96-well Plate
Bacterial Culture (E. coli, High-copy Plasmids)
Elution Volume: 100 - 200 μL
Centrifuge, Vacuum manifold (e.g. NucleoVac 96)
Filter Plates, Binding Plates, Buffers, RNase A
Core Kit
Up to 5 mL
45 min/plate
Room temperature (18 - 25 °C)
Silica Membrane and Endotoxin-reduction Technology
Binding capacity: 20 μg
Transfection-grade plasmid DNA
Rapid purification of plasmids for transfection
4 x 96
A260/A280: 1.80 - 1.85
< 15 kbp
5 - 20 μg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • NucleoSpin 96 Plasmid T.-gr. Core (4x96)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide, dodecyl sulphate, guanidine hydrochloride,triethanolamine, 2-propanol, RNAse

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.