missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Nucleospin™ 96 Plasmid Transfection-grade
A fast way to purify transfection-grade plasmid DNA using mini-format technology to reduce endotoxins from bacterial lysates. Full and Core kits utilize a 96-well plate format for an efficient high-throughput process.
£990.00 - £4825.00
Specifications
Bacterial Endotoxins | <50 EU/ng DNA |
---|---|
Content And Storage | Room temperature (18 - 25 °C) |
Format | 96-well Plate |
Isolation Technology | Silica Membrane and Endotoxin-reduction Technology |
Sample Type | Bacterial Culture (E. coli, High-copy Plasmids) |
Product Code | Brand | No. of Samples | Product Type | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Samples | Product Type | Includes | Price | Quantity & Availability | |||
15553636
|
Macherey-Nagel™
740492.4 |
4 x 96 | Core Kit | Filter Plates, Binding Plates, Buffers, RNase A |
£1300.00
384 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15543636
|
Macherey-Nagel™
740492.24 |
24 x 96 | Core Kit | Filter Plates, Binding Plates, Buffers, RNase A |
£4825.00
2304 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15533636
|
Macherey-Nagel™
740491.4 |
4 x 96 | Full Kit | Filter Plates, Binding Plates, MN Wash Plates, Culture Plates, Elution Plates, Buffers, RNase A |
£990.00
384 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Endotoxins are co-purified during typical plasmid preparations from bacterial lysates. Since they interfere with eukaryotic cell survival, endotoxin reduction is essential prior to cell transfection. NucleoSpin™ Plasmid Transfection-grade kits are designed to reduce endotoxins from bacterial lysates.
- Purifies high-copy plasmids for transfection
- Novel technology to diminish endotoxin content (< 50 EU/μg DNA) for successful transfection
- Rapid purification from E. coli via automatic, manual, vacuum or centrifugation processing
- Ultra-fast procedure simplifies the work-flow
- High-throughput — 96-well plates allow for easy automation and supports plasmid screening
- Downstream applications: transfection of common cell lines (lipofection, electroporation)
- Also suitable for standard molecular biology application, such as cloning and sequencing
Specifications
<50 EU/ng DNA | |
96-well Plate | |
Bacterial Culture (E. coli, High-copy Plasmids) | |
Elution Volume: 100 - 200 μL | |
Centrifuge, Vacuum manifold (e.g. NucleoVac 96) | |
A260/A280: 1.80 - 1.85 | |
< 15 kbp | |
5 - 20 μg |
Room temperature (18 - 25 °C) | |
Silica Membrane and Endotoxin-reduction Technology | |
Binding capacity: 20 μg | |
Transfection-grade plasmid DNA | |
Rapid purification of plasmids for transfection | |
Up to 5 mL | |
45 min/plate |
Safety and Handling
missing translation for 'shelfLife' : At least 1 year
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- NucleoSpin 96 Plasmid T.-gr. Core (4x96)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide, dodecyl sulphate, guanidine hydrochloride,triethanolamine, 2-propanol, RNAse