missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ miRNA Kit

Isolate small and large RNA in parallel. NucleoSpin™ miRNA Kits work on a wide varitey of tissue and cell culture formats.

Brand:  Macherey-Nagel™ 740971.50

303.23 GBP valid until 2024-12-31
Use promo code "23283" to get your promotional price.



Code : 86

Additional Details : Weight : 0.20500kg Transport : UN number : 1993 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 12798622

  • £479.88 / 50 reactions
Estimated Shipment: 08-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Features
  • Parallel isolation of small and large RNA
  • RNA purification fractionated by size: 
  • Isolation of small RNA only (< 200 b),
  • Isolation of small RNA (< 200 b) and large RNA (> 200 b) in two separate fractions
  • Isolation of total RNA (small and large RNA in one fraction)
  • Additional isolation of total protein fraction ready to use for SDS-PAGE and Western blot analysis
  • Excellent RNA recovery and purity by chaotropic salt lysis without phenol/chloroform 
  •   (patent pending)
  • NucleoSpin™ Filters for efficient sample homogenization
  • rDNase for efficient on-column removal of genomic DNA
Applications
  • Parallel isolation of small and large RNA from human / animal tissue and cultured cells, from plant tissue,
  •    and in combination with phenol / chloroform (e.g., TRIzol™) lysis 
  • Purification of siRNA and large dsRNA from DICER reactions
  • Typical downstream applications: real-time RT-PCR, Northern blotting, chip hybridization
Specifications

Specifications

Mini Spin Column
NucleoSpin™ RNA Columns, NucleoSpin™ miRNA Columns, NucleoSpin™ Protein Removal Columns, Collection Tubes (2 mL, 2 mL lid, 1.5 mL), NucleoSpin™ Filters, buffers, RNase-free rDNase
NucleoSpin™ miRNA Kit
Small RNA: < 200 nt, large RNA: > 200 nt
10μg small RNA, 95μg large RNA
Purification of siRNA and large dsRNA from DICER reactions
50
Yes
<45 min./6 Reaction and <35 min./6 Reaction
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers