missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ RNA Virus 50 Preps
Mini spin kit for the isolation of viral RNA and DNA from cell-free biological fluids
Brand: Macherey-Nagel™ 740956.50
177.12 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
NucleoSpin™ RNA Virus is designed for the isolation of viral nucleic acids from serum, plasma or any cell-free biological fluids, featuring a special type of membrane which has a high binding capacity for nucleic acids.The procedure is rapid and easy. The lysate is incubated for 5 to 10min and applied to the NucleoSpin™ column. After two washing steps to remove contaminations the RNA can be eluted with water or low-salt buffer. The kit contains all necessary buffers, spin columns and detailed protocols. Generally, NucleoSpin™ RNA Virus is suited for the parallel purification of viral RNA and DNA (support protocol). For example, isolation of HBV DNA is performed with an additional proteinase K digest (enzyme not included in the kit). For isolation of viral DNA with highest sensitivities, the NucleoSpin™ Blood kit is recommended.
Kit components: NucleoSpin™ RNA Virus columns, 2mL collecting tubes, buffers, RNase-free water, carrier RNA.
• Silica membrane technology
• Typical recovery: >90%
• Sample material: 150μL serum, plasma, cell-free biological fluids
• Fragment size: 100bp to 30kb
• Elution volume: 50μL
• Binding capacity: 40μg
• Preparation time: 30min/4-6 preps
• Format: mini spin column
• Support protocol for parallel isolation of viral RNA and DNA (proteinase K required)
• Applicable for HCV, HBV, HAV, HIV, HSV, HPV, VZV, EBV, parvovirus B19, H5N1
Specifications
18°C to 25°C | |
Cell-free Biological Fluids, Plasma, Serum | |
50 Preps | |
100 bp−approx. 50 kbp |
Mini Prep | |
Isolation of Viral Nucleic Acids | |
< 150 L | |
Viral RNA and DNA |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash with water thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
Supplemental information
- MIXTURE LIST-Contains : guanidine hydrochloride, ethanol