Learn More
NUDT13 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25912-1-AP-150UL
This item is not returnable.
View return policy
Description
The NUDT13 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human NUDT13. This antibody recognizes human antigen. The NUDT13 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
NUDT13 | |
Polyclonal | |
Rabbit | |
Primary | |
Human |
Western Blot, ELISA | |
Unconjugated | |
150 μL | |
25961 | |
IgG |