missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT13 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | NUDT13 |
---|---|
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17422333
|
Proteintech
25912-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17412333
|
Proteintech
25912-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The NUDT13 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human NUDT13. This antibody recognizes human antigen. The NUDT13 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
NUDT13 | |
Polyclonal | |
Rabbit | |
25961 | |
Primary |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG |