missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates

Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.

Brand:  Thermo Scientific™ 267407

170.00 GBP valid until 2024-12-14
Use promo code "21632" to get your promotional price.


 View more versions of this product

Product Code. 10355661

  • £255.00 / Pack of 120
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price

This item is not returnable. View return policy

Description

Description

Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.

Features:

  • Shared-wall design increases the well volume
  • Wider wells and rounded bottom improve mixing
  • Conical bottom well for optimal sample recovery
  • Black and white plates available for fluorescent- and luminescent-based applications
  • Low binding polypropylene for homogeneous assays and storage
TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Barcode
96-well
96-well Polypropylene Sample Processing & Storage Plates
Autoclavable
Low binding PP
Yellow
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
500 μL
450 μL
Non-treated
Yes
120
120/Cs.
Yes
ANSI/ SLAS Standard
Certified RNase and DNase Free. ANSI Compliant.
Without Lid
8 x 12
10 pk., 120 cs.
Nunc
96-well plate
Polypropylene
96
Non-sterile
Round
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
10
For Research Use Only. Not for use in diagnostic procedures.
Certified Quality
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers