Learn More
Thermo Scientific™ Nunc™ EasYDish™ Dishes with Nunclon™ Supra Surface
Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.
Brand: Thermo Scientific™ 150470
234.50 GBP valid until 2025-12-31
Use promo code "24071" to get your promotional price.
Description
Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The beveled grip ring is easy to hold and stays secure in your gloved hand. Available in four sizes: 35, 60, 100, and 150 mm. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.
Features of Nunc EasYDish Dishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

Specifications
Not Autoclavable | |
Clear | |
Single-use | |
Polystyrene | |
Sterile | |
500 Dishes | |
13 mm |
Flat | |
8.8 cm2 | |
Not Gridded | |
Round | |
Cell Culture Dish | |
10 Dishes | |
35 mm |
Special Offers
-
Thermo Scientific Nunc
,
Thermo Scientific Nalgene
,
Gibco
,
Invitrogen
,
Thermo Scientific
Cell Biology Promotion
Discover how you can maximise your savings and boost your research to the next level with our top-of-the-line Gibco and Thermo Scientific products.Promo Code: 24071Expires: 2025-06-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Laboratory Use