missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ EasYDish™ Dishes with Nunclon™ Supra Surface

Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.

£185.00 - £335.00

Specifications

Autoclavable Not Autoclavable
Bottom Shape Flat
Color Clear
Disposable Single-use
Gridded Not Gridded
View More Specs

Products 5
Product Code Brand Diameter (Metric) Culture Area Height (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Culture Area Height (Metric) Price Quantity & Availability  
17947422
View Documents Promotion Details
Thermo Scientific™
150470
35 mm 8.8 cm2 13 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17987422
View Documents Promotion Details
Thermo Scientific™
150472
60 mm 21.5 cm2 16 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17977422
View Documents Promotion Details
Thermo Scientific™
150476
100 mm 56.7 cm2 21 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17907432
View Documents Promotion Details
Thermo Scientific™
150474
100 mm 56.7 cm2 17 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17967422
View Documents Promotion Details
Thermo Scientific™
150478
150 mm 145 cm2 21 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The beveled grip ring is easy to hold and stays secure in your gloved hand. Available in four sizes: 35, 60, 100, and 150 mm. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc EasYDish Dishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

Specifications

Specifications

Not Autoclavable
Clear
Not Gridded
Round
Cell Culture Dish
Flat
Single-use
Polystyrene
Sterile
10 Dishes
Videos
SDS
Documents

Documents

Product Certifications

For Laboratory Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ EasYDish™ Dishes with Nunclon™ Supra Surface

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.