Learn More
Thermo Scientific™ Nunc™ IVF Dishes

Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.
£689.00
Specifications
Certifications/Compliance | CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested |
---|---|
Material | Polystyrene (USP Class VI) |
Sterility | Sterile |
Surface Treatment | Non-treated |
No. per Pack | 10 |
Product Code | Brand | Diameter (Metric) | Height (Metric) | Culture Area | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Height (Metric) | Culture Area | Price | Quantity & Availability | |||
10170753
![]() |
Thermo Scientific™
150255 |
35 mm | 11 mm | 8.8 cm2 |
£689.00
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.
Product features for reliable results:
- Facilitate gas exchange with vented lid
- Available in 35mm, 60mm and 90mm sizes
- Certificates of conformity available for every lot
Rigorously tested, certified and validated for IVF:
- CE-marked (Notified Body 2460), FDA-listed
- Produced under ISO 13485 with full lot traceability
- Sterile (SAL 10-6)
- Non-pyrogenic (LAL tested)
- USP Class VI polystyrene
- Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)
Warranty: 90 days
Specifications
CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested | |
Sterile | |
10 |
Polystyrene (USP Class VI) | |
Non-treated |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.