Learn More
Thermo Scientific™ Nunc™ MicroWell™ MiniTrays
Create more space in lab freezers and inubators with Nunc MicroWell MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
Brand: Thermo Scientific™ 439225
224.05 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 76
Additional Details : Weight : 1.71000kg
Description
Create more space in lab freezers and inubators with Thermo Scientific™ Nunc™ MicroWell™ MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
- Limited adsorption of proteins
- Lids have keyed tabs for proper orientation
- Large raised alphanumeric coordinates
- Unique surface treatment to maximize sample and reagent mixing
- MiniTrays for serological applications have slightly smaller well dimensions
Specifications
Closure Included | |
MicroWell MiniTray for serological applications | |
Polystyrene | |
Non-sterile | |
Cell Culture Dish | |
For serological applications | |
100 | |
84 mm | |
59 mm |
Polystyrene | |
Flat | |
Rectangular | |
Nunclon Delta | |
60 | |
Lid | |
10 | |
59 mm |