missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ MicroWell™ MiniTrays
Create more space in lab freezers and inubators with Nunc MicroWell MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
Brand: Thermo Scientific™ 472400
585.00 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Create more space in lab freezers and inubators with Thermo Scientific™ Nunc™ MicroWell™ MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
- Limited adsorption of proteins
- Lids have keyed tabs for proper orientation
- Large raised alphanumeric coordinates
- Unique surface treatment to maximize sample and reagent mixing
- MiniTrays for serological applications have slightly smaller well dimensions
Specifications
Closure Not Included | |
Flat | |
Rectangular | |
Nunclon Delta | |
60 | |
400 | |
84 mm | |
59 mm |
MicroWell MiniTray | |
Polystyrene | |
Sterile | |
Cell Culture Dish | |
S Plate | |
100 | |
59 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction