Learn More
Thermo Scientific™ Nunc™ Multidishes with UpCell™ Surface

Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins.
Brand: Thermo Scientific™ 174899
194.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with the temperature-responsive surface of Thermo Scientific™ Nunc™ UpCell™ Dishes. Simply transfer the culture to room temperature to collect cells in suspension or in the form of cell sheet with the help of a supportive membrane.
Traditional cell harvesting by enzymatic and mechanical methods often compromises the integrity of surface proteins and the viability of harvested cells. By contrast, UpCell Surface allows cell harvesting by simply reducing the temperature of the cell culture to below 32°C without the need of enzyme, resulting in highly viable cells with intact cell surface proteins.- No trypsinization—preserve cell surface proteins
- No physical force—get high cell viability
- For culture passaging, single-cell analyses and cell transplantation research
- 6-well multidish comes with supportive membranes that enable harvesting of cell sheets and creations of 3-D tissue models
- Minimal hands-on time
- Customized bar coding is available
Recommended for:
Culture passaging; Single-cell analyses, Cell transplantation research, 3D tissue models

Specifications
24 well, UpCell | |
1 mL (Well) | |
Multi Dish | |
6 | |
Cell Culture-Treated | |
24 |
1.9 sq. cm | |
1 mL | |
1 | |
Clear | |
Sterile | |
Rectangular, Flat Bottom (F-well) |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
NU24W2439 | Certificate of Compliance | 07/03/2025 | 174899 |
NU24W2434 | Certificate of Compliance | 14/12/2024 | 174899 |
NU24W2422 | Certificate of Compliance | 06/12/2024 | 174899 |
NU24W2419 | Certificate of Compliance | 06/12/2024 | 174899 |
NU24W2411 | Certificate of Compliance | 06/12/2024 | 174899 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. For research and single-use only