Learn More
Thermo Scientific™ Nunc™ Multidishes with UpCell™ Surface
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins.
Brand: Thermo Scientific™ 174901
198.90 GBP valid until 2024-12-31
Use promo code "21928" to get your promotional price.
Description
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with the temperature-responsive surface of Thermo Scientific™ Nunc™ UpCell™ Dishes. Simply transfer the culture to room temperature to collect cells in suspension or in the form of cell sheet with the help of a supportive membrane.
Traditional cell harvesting by enzymatic and mechanical methods often compromises the integrity of surface proteins and the viability of harvested cells. By contrast, UpCell Surface allows cell harvesting by simply reducing the temperature of the cell culture to below 32°C without the need of enzyme, resulting in highly viable cells with intact cell surface proteins.- No trypsinization—preserve cell surface proteins
- No physical force—get high cell viability
- For culture passaging, single-cell analyses and cell transplantation research
- 6-well multidish comes with supportive membranes that enable harvesting of cell sheets and creations of 3-D tissue models
- Minimal hands-on time
- Customized bar coding is available
Recommended for:
Culture passaging; Single-cell analyses, Cell transplantation research, 3D tissue models
Specifications
6 well, UpCell | |
3 mL (Well) | |
Multi Dish | |
6 | |
UpCell, Temperature Responsive | |
6 |
9.6 sq. cm | |
3 mL | |
1 | |
Clear | |
Sterile | |
Rectangular, Flat Bottom (F-well) |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. For research and single-use only