missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
GREENER_CHOICE

Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.

Marca:  Thermo Scientific™ 236370

84.43 GBP valid until 2025-06-30
Use promo code "25075" to get your promotional price.


 vedi altre versioni di questo prodotto

Codice prodotto. 11948231

  • £189.00 / Pack of 100
In Stock
to see stock.
Edge
Esplora altre offerte speciali
Aggiungi al carrello

Descrizione

Descrizione

Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.

  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
TRUSTED_SUSTAINABILITY
Specifica

Specifica

Clear
SEALING TAPE, POLYESTER NS
Non-sterile
96
100
General
Nunc
No
-40°C to +120°C
All Microplates
Yes
Life Science Reagent Resistant
Polyester
Not Autoclavable
100
General purpose, sample detection
Adhesive Seal
Acrylic
No
All well shapes
General
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

  • Multiple Manufacturers
    Save up to 94% on Teaching Lab Essentials
    Promo Code25721

    Expires2025-05-01

    New Offer
    Save Now
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
  • Thermo Scientific Matrix
    Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.
    Promo Code: 25075

    Expires2025-06-30

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils > Clear polyester adhesive sealing tape, 100/100

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato