missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NVP ADW 742
ATP-competitive inhibitor of IGF1R
£140.00 - £533.00
Specifications
Purity | >99% |
---|---|
Product Type | NVP ADW 742 |
Molecular Weight (g/mol) | 453.58 |
Inhibitors | NVP ADW 742 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753968
|
Tocris Bioscience™
5247/10 |
10 mg |
£140.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18743158
|
Tocris Bioscience™
5247/50 |
50 mg |
£533.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATP-competitive inhibitor of IGF1R (IC50 in the range of 0.1-0.2 μM). Also inhibits c-Kit kinase activity (IC50 in the range of 3-5 μM). Exhibits a synergistic effect with Imatinib, enhancing the sensitivity of multiple small cell lung cancer cell lines to the chemotherapeutics etoposide (Cat. No. 1226) and carboplatin (Cat. No. 2626).Specifications
>99% | |
NVP ADW 742 | |
453.58 | |
NVP ADW 742 |
For Research Use Only