Learn More
Masterflex™ Nylon Straight Barbed Union
Brand: Masterflex™ 30612-30
This item is not returnable.
View return policy
Specifications
Straight Barbed Union | |
Nylon | |
-46°C | |
121°C |
19.05 mm | |
19.05mm | |
10 Pack |