missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals O-Methyl-L-tyrosine, 98%
£29.50 - £241.00
Chemical Identifiers
CAS | 6230-11-1 |
---|---|
Molecular Formula | C10H13NO3 |
Molecular Weight (g/mol) | 195.22 |
MDL Number | MFCD00002604 |
InChI Key | GEYBMYRBIABFTA-IMWMWJONNA-N |
Synonym | 4-methoxy-l-phenylalanine, o-methyl-l-tyrosine, h-tyr me-oh, o-methyltyrosine, s-2-amino-3-4-methoxyphenyl propanoic acid, p-methoxy-l-phenylalanine, 2s-2-amino-3-4-methoxyphenyl propanoic acid, tyrosine, o-methyl, l-4-methoxyphenylalanine, h-4-methoxy-phe-oh |
PubChem CID | 2723935 |
IUPAC Name | (2S)-2-amino-3-(4-methoxyphenyl)propanoic acid |
SMILES | COC1=CC=C(C[C@H](N)C(O)=O)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10433035
|
Thermo Scientific Chemicals
169752500 |
250 mg | Glass Bottle |
£29.50
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10042150
|
Thermo Scientific Chemicals
169750010 |
1 g | Glass Bottle |
£79.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10167703
|
Thermo Scientific Chemicals
169750050 |
5 g | Glass Bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6230-11-1 | |
195.22 | |
GEYBMYRBIABFTA-IMWMWJONNA-N | |
2723935 | |
COC1=CC=C(C[C@H](N)C(O)=O)C=C1 |
C10H13NO3 | |
MFCD00002604 | |
4-methoxy-l-phenylalanine, o-methyl-l-tyrosine, h-tyr me-oh, o-methyltyrosine, s-2-amino-3-4-methoxyphenyl propanoic acid, p-methoxy-l-phenylalanine, 2s-2-amino-3-4-methoxyphenyl propanoic acid, tyrosine, o-methyl, l-4-methoxyphenylalanine, h-4-methoxy-phe-oh | |
(2S)-2-amino-3-(4-methoxyphenyl)propanoic acid |
Specifications
6230-11-1 | |
100.0 | |
98% | |
Authentic | |
MFCD00002604 | |
4-methoxy-l-phenylalanine, o-methyl-l-tyrosine, h-tyr me-oh, o-methyltyrosine, s-2-amino-3-4-methoxyphenyl propanoic acid, p-methoxy-l-phenylalanine, 2s-2-amino-3-4-methoxyphenyl propanoic acid, tyrosine, o-methyl, l-4-methoxyphenylalanine, h-4-methoxy-phe-oh | |
COC1=CC=C(C[C@H](N)C(O)=O)C=C1 | |
-7° to -9° (20°C, 589nm) (c=2, 1N HCl) | |
2723935 | |
≥97.5% | |
Powder |
97.5 | |
Beige to White | |
C10H13NO3 | |
CH3OC6H4CH2CH(NH2)CO2H | |
14, I, 668 | |
GEYBMYRBIABFTA-IMWMWJONNA-N | |
(2S)-2-amino-3-(4-methoxyphenyl)propanoic acid | |
195.22 | |
195.22 | |
−8° (20°C c=2,1N HCl) | |
O-Methyl-L-tyrosine |
RUO – Research Use Only