Learn More
DWK Life Sciences O-ring, made of nitrile, for safety standard ground sockets
Component of the DURAN™ safety ground joint.
Brand: DWK Life Sciences 286217508
This item is not returnable.
View return policy
Description
Nitrile O-rings for DURAN™ safety ground joints together with the screw cap secure the connection between core and sleeve.
- In combination with the screw cap, the safety ground joints (core and sleeve) can be sealed tightly.
Specifications
To secure the security grindings; Sealing | |
NS 34/35 | |
10/Pk. | |
Nitrile |