Learn More
DWK Life Sciences O-ring, made of nitrile, for safety standard ground sockets
Component of the DURAN™ safety ground joint.
£5.00 - £12.60
Specifications
Material | Nitrile |
---|---|
For Use With (Application) | To secure the security grindings; Sealing |
Quantity | 10/Pk. |
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
15921561
|
DWK Life Sciences
286217705 |
NS 45/40 |
£12.60
Pack of 10 |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15864608
|
DWK Life Sciences
286216803 |
NS 14/23 |
£5.00
Pack of 10 |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
15991551
|
DWK Life Sciences
286217002 |
NS 19/26 |
£7.45
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15884608
|
DWK Life Sciences
286217405 |
NS 29/32 |
£9.85
Pack of 10 |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
15894608
|
DWK Life Sciences
286217508 |
NS 34/35 |
£10.50
Pack of 10 |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
15911561
|
DWK Life Sciences
286217208 |
NS 24/29 |
£9.25
Pack of 10 |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
Description
Nitrile O-rings for DURAN™ safety ground joints together with the screw cap secure the connection between core and sleeve.
- In combination with the screw cap, the safety ground joints (core and sleeve) can be sealed tightly.
Specifications
Nitrile | |
To secure the security grindings; Sealing | |
10/Pk. |