missing translation for 'onlineSavingsMsg'
Learn More
pH Calibration Buffer Bottles, Oakton™ Product Code.: 11710069
compliance-icons

pH Calibration Buffer Bottles, Oakton™

Product Code. 11710069 Shop All Cole-Parmer Products
12.46
Click to view available options
pH:
1.68
7.00
10.00
12.46
This item is not returnable. View return policy

Product Code. 11710069

Brand: Cole Parmer 0065412

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use these PH buffers for meters with 0.01 resolution. Oakton™ pH Calibration Buffer Bottles are standardized against NIST-traceable pH references within 0.01 pH at 77°F (25°C).

  • pH buffer solutions allow the user to adjust their system to read accurate measurements
  • Store buffers away from direct sunlight.

Order Info

Shipping weight: 1-1/4 lbs. (0.6kg).

Certifications

NIST traceable, Certified to NIST traceable standards.

Specifications

For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 12.46
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
pH Calibration Buffer Bottles, Oakton™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.