missing translation for 'onlineSavingsMsg'
Learn More
pH Calibration Buffer Bottles, Oakton™ Product Code.: 11739679
compliance-icons

pH Calibration Buffer Bottles, Oakton™

Product Code. 11739679 Shop All Cole-Parmer Products
1.68
Click to view available options
pH:
1.68
7.00
10.00
12.46
This item is not returnable. View return policy

Product Code. 11739679

Brand: Cole Parmer 0065401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use these PH buffers for meters with 0.01 resolution. Oakton™ pH Calibration Buffer Bottles are standardized against NIST-traceable pH references within 0.01 pH at 77°F (25°C).

  • pH buffer solutions allow the user to adjust their system to read accurate measurements
  • Store buffers away from direct sunlight.

Order Info

Shipping weight: 1-1/4 lbs. (0.6kg).

Certifications

NIST traceable, Certified to NIST traceable standards.

Specifications

For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 1.68
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
compliance-icons
Product Identifier
  • pH 1.68 Buffer
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P202-Do not handle until all safety precautions have been read and understood.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
pH Calibration Buffer Bottles, Oakton™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.