missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ OB 24 hydrochloride
Potent and selective heme oxygenase 1 (HO-1) inhibitor
£171.00 - £640.00
Specifications
Chemical Name or Material | 1-[[2-[2-(4-Bromophenyl)ethyl]-1,3-dioxolan-2-yl]methyl]-1H-imidazole hydrochloride |
---|---|
CAS | 939825-12-4 |
Assay Percent Range | >98% |
Molecular Formula | C15H17BrN2O2.HCl |
Formula Weight | Observed MW: 373.67 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785696
|
Tocris Bioscience™
6119/10 |
10 mg |
£171.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18745456
|
Tocris Bioscience™
6119/50 |
50 mg |
£640.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective heme oxygenase 1 (HO-1) inhibitor (IC50 values are 1.9 and >100 μM for HO-1 and HO-2 respectively), reduces protein carbonylation and reactive oxygen species formation in advanced prostate cancer cells (PCA). Inhibits cell proliferation in vitro and PCA tumor growth and lymph node/lung metastases in vivo. Synergizes with Taxol (Cat. No. 1097).Specifications
1-[[2-[2-(4-Bromophenyl)ethyl]-1,3-dioxolan-2-yl]methyl]-1H-imidazole hydrochloride | |
>98% | |
Observed MW: 373.67 |
939825-12-4 | |
C15H17BrN2O2.HCl |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ OB 24 hydrochloride