Learn More
Tocris Bioscience™ OB 24 hydrochloride
Potent and selective heme oxygenase 1 (HO-1) inhibitor
Brand:  Tocris Bioscience™ 6119/50
 View more versions of this product
Product Code. 18745456
  • £640.00 / 50mg
Explore more special offers
Description
Description
OB 24 hydrochloride is a potent and selective heme oxygenase 1 (HO-1) inhibitor (IC50 values are 1.9 and >100 μM for HO-1 and HO-2 respectively), reduces protein carbonylation and reactive oxygen species formation in advanced prostate cancer cells (PCA). Inhibits cell proliferation in vitro and PCA tumor growth and lymph node/lung metastases in vivo. Synergizes with Taxol (Cat. No. 1097).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1-[[2-[2-(4-Bromophenyl)ethyl]-1,3-dioxolan-2-yl]methyl]-1H-imidazole hydrochloride
>98%
C15H17BrN2O2.HCl
939825-12-4
50 mg
Observed MW: 373.67
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tocris Bioscience™ OB 24 hydrochloride > 50mg