Learn More
Octadecyltrimethoxysilane, 90%, Tech., Thermo Scientific Chemicals
£88.54 - £278.39
Chemical Identifiers
CAS | 3069-42-9 |
---|---|
Molecular Formula | C21H46O3Si |
Molecular Weight (g/mol) | 374.69 |
MDL Number | MFCD00043060 |
InChI Key | SLYCYWCVSGPDFR-UHFFFAOYSA-N |
Synonym | octadecyltrimethoxysilane, trimethoxy octadecyl silane, n-octadecyltrimethoxysilane, stearyltrimethoxysilane, silane, trimethoxyoctadecyl, octadecyl-trimethoxysilane, acmc-209hi7, ksc491g4l, trimethoxy octadecyl silane, technical grade |
PubChem CID | 76486 |
IUPAC Name | trimethoxy(octadecyl)silane |
SMILES | CCCCCCCCCCCCCCCCCC[Si](OC)(OC)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10657251
|
Thermo Scientific Acros
313600250 |
25 mL | Glass bottle |
£88.54
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10300994
|
Thermo Scientific Acros
313601000 |
100 mL | Glass bottle |
£278.39
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3069-42-9 | |
374.69 | |
SLYCYWCVSGPDFR-UHFFFAOYSA-N | |
76486 | |
CCCCCCCCCCCCCCCCCC[Si](OC)(OC)OC |
C21H46O3Si | |
MFCD00043060 | |
octadecyltrimethoxysilane, trimethoxy octadecyl silane, n-octadecyltrimethoxysilane, stearyltrimethoxysilane, silane, trimethoxyoctadecyl, octadecyl-trimethoxysilane, acmc-209hi7, ksc491g4l, trimethoxy octadecyl silane, technical grade | |
trimethoxy(octadecyl)silane |
Specifications
3069-42-9 | |
100.0 | |
Colorless to Yellow | |
170°C (0.1 mmHg) | |
Authentic | |
C21H46O3Si | |
1.4380 to 1.44 | |
0.88 | |
Solubility in water: insoluble (reacts) | |
CCCCCCCCCCCCCCCCCC[Si](OC)(OC)OC | |
374.69 | |
374.69 | |
Technical | |
Octadecyltrimethoxysilane |
88.0 | |
13°C to 17°C | |
0.8800g/mL | |
139°C | |
88% min. Total octadecyltrimethoxysilanes (GC) | |
CH3(CH2)17Si(OCH3)3 | |
MFCD00043060 | |
octadecyltrimethoxysilane, trimethoxy octadecyl silane, n-octadecyltrimethoxysilane, stearyltrimethoxysilane, silane, trimethoxyoctadecyl, octadecyl-trimethoxysilane, acmc-209hi7, ksc491g4l, trimethoxy octadecyl silane, technical grade | |
SLYCYWCVSGPDFR-UHFFFAOYSA-N | |
trimethoxy(octadecyl)silane | |
76486 | |
90% | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 221-339-2
Recommended Storage : Product may become turbid on storage
RUO – Research Use Only