Learn More
Octanoyl chloride, 99%, Thermo Scientific Chemicals
£11.70 - £243.00
Chemical Identifiers
CAS | 111-64-8 |
---|---|
MDL Number | MFCD00000766 |
InChI Key | REEZZSHJLXOIHL-UHFFFAOYSA-N |
Synonym | capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride |
PubChem CID | 8124 |
IUPAC Name | octanoyl chloride |
SMILES | CCCCCCCC(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10697092
|
Thermo Scientific Acros
129430050 |
5 g | Glass bottle | ||||||
10607292
|
Thermo Scientific Acros
129431000 |
100 g | Glass bottle |
£17.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10489050
|
Thermo Scientific Acros
129435000 |
500 g | Glass bottle |
£52.20
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10249583
|
Thermo Scientific Acros
129430025 |
2.5 kg | Glass bottle |
£243.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-64-8 | |
REEZZSHJLXOIHL-UHFFFAOYSA-N | |
8124 | |
CCCCCCCC(=O)Cl |
MFCD00000766 | |
capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride | |
octanoyl chloride |
Specifications
1% max. | |
98.5 | |
0.9500g/mL | |
99% | |
CH3(CH2)6COCl | |
MFCD00000766 | |
capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride | |
CCCCCCCC(=O)Cl | |
162.657 | |
162.66 | |
Liquid |
111-64-8 | |
100.0 | |
Authentic | |
C8H15ClO | |
1.4340 to 1.4360 (20°C, 589nm) | |
02, 348 | |
REEZZSHJLXOIHL-UHFFFAOYSA-N | |
octanoyl chloride | |
8124 | |
≥98.5% | |
Octanoyl chloride, 99% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to
Danger
missing translation for 'einecsNumber' : 203-891-6
missing translation for 'rtecsNumber' : RH1570000
missing translation for 'tsca' : TSCA
RUO – Research Use Only