Learn More
3M™ Oil Sorbent Boom
A double boom with two, 4 inch scrims encased tubes of particulate sorbent material, secured by a UV resistant netting.
Brand:  3M™ 7000088226
 View more versions of this product
Product Code. 17108650
  • £534.00 / Pack of 2
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used to cover large spills, or as a precaution in areas where large spills may occur
  • Ideal for absorbing oil from areas where water may also be present
  • Link sorbent booms together by built-in connectors to deploy over large areas
  • UV-resistant to prevent deterioration and disintegration
  • Designed for long-term use in ditches, streams, rivers, lakes, docks, and piers
  • Cost-effective as oil, not water, is recovered
  • Can be used outside or in-plant
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sorbent Boom
200 mm
5 m
Brown
Workplace Safety, Spill Prevention
T-270 GA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Oil Sorbent Boom > 5 m, 200 mm