Learn More
Thermo Scientific™ OLIGO™ R3 Reversed - Phase Resin
Thermo Scientific™ Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules.
Brand: Thermo Scientific™ 1133907
Additional Details : Weight : 0.58290kg
Description
Thermo Scientific™ Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules. The combination of 30-μm particle size, resistance to high pH, and high-phase ratio, makes Oligo R3 resin particularly suited to preparative purification of synthetic oligonucleotides and peptides. The packing consists of cross-linked poly-divinylbenzene particles. Oligo R3 bulk resin differs somewhat in selectivity from conventional silica-based, reversed-phase resins and is similar to high-carbon-loading through C18 supports. In addition, the complete lack of residual silanol activity may modify the selectivity of ionic species.
Features of Oligo R3 resin and pre-packed columns include:
- High performance chromatography resin for analytical to process scale separations
- High resolution preparative purification at high flow rates
- Chemical stability—allows aggressive cleaning and sanitization
- Poly-divinylbenzene particles—yield a robust, easily packable matrix
- Combines low back pressure, high pH stability, and high capacity
Specifications
1 L | |
R3 Reversed-Phase Resin | |
Store at room temperature. | |
R3 |
OLIGO, POROS | |
30 μm | |
Room Temperature | |
Bulk Media |